Learn More
3M™ PELTOR™ Welding Ear Muff, Behind-the-Head
Click to view available options
Welding Helmet Ear Muff
Description
- H505B ear muff low-profile design fits comfortably under most welding helmets particularly 3M™ Speedglas™ 9100 Series
- Wire headband offers comfortability during prolonged use
- Ear cups tilt to ensure customized fit
- Two-point suspension for balanced pressure distribution
- Semi-soft cup surface minimizes scratch sound transmission
Compliance
3M™ products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications
Specifications
ANSI S3.1974 | Yes |
CSA Z94.2 | Class B |
Cushion Color | Black |
For Use With (Equipment) | 3 M™ Speedglas™ 9100 Series and Other Welding Helmets |
Headband Material | Wire |
Headband Cushion | No |
For Use With (Application) | Grinding, Machine Operations, Welding |
Model | H505B |
Noise Reduction Rating (NRR) | 17 dB |
Headband Color | Silver |
Show More |
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
3M™ PELTOR™ Welding Ear Muff, Behind-the-Head >
Spot an opportunity for improvement?Share a Content Correction