Learn More
3M™ Rugged Comfort 6500 Series Quick Latch Half Facepiece Reusable Respirator Product Code.: 15757707
Estimated Shipment: 01-08-2025
Only null left
Add to basket
3M™ Rugged Comfort 6500 Series Quick Latch Half Facepiece Reusable Respirator
Product Code. 15757707
Medium, Each
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.
Product Code. 15757707
Brand: 3M™ 7000128239
Only null left
Estimated Shipment: 01-08-2025
Add to basket
Only null left
Add to basket
You may only order up to the available stock. For more details call customer services on +45 70 27 99 20.
Rugged Comfort reusable respirator combines comfort, durability, and stability with an added quick latch mechanism to easily enable the mask to drop down from face without removing head straps.
  • Resilient silicone faceseal offers extended facepiece life even in high-heat environments
  • Quick latch design eliminates need to remove a hardhat or faceshield when lowering or raising the mask
  • Sleek, low-profile design
  • Special valve cover helps to direct exhaled breath and moisture downward
  • 3M’s proprietary Cool Flow™ Valve helps to make breathing easier by reducing heat and moisture
  • Fits securely and comfortably on the head
  • Compatible with all 3M™ bayonet-style filters and cartridges:
    • 6000 Series Cartridges
    • 2000, 2200, 7000 or 5000 Series Filters
TRUSTED_SUSTAINABILITY
Specifications
Certifications (NIOSH) NIOSH 42 CFR Part 84
Certifications (OSHA) OSHA 29 CFR Part 1920.134
Connection Type Bayonet
Color Gray, Teal
Product Type Half-Mask Respirator
Size Medium
Facepiece Material Silicone
For Use With (Application) Facility maintenance, grinding, machine operations, sanding, welding, demolition, assembly, cleaning
For Use With (Equipment) Line Compressed Air System SA Series, 3M™ 6000 Series cartridges and 2000, 2200, 7000 or 5000 Series filters
Includes Quick Latch
Model 6502QL
Suspension Type Yolk and cradle
Weight (English) 0.37 lb.
Weight (Metric) 0.16 kg
Strap Attachment Type Quick Latch Mechanism
Show More Show Less
Product Content Correction
Product Title
3M™ Rugged Comfort 6500 Series Quick Latch Half Facepiece Reusable Respirator >