Learn More
3M™ Versaflo™ TR-600/800 PAPR Accessory, Cleaning and Storage Kit TR-653 / 37373 (AAD)
Click to view available options
:
Cleaning and Storage Kit TR-653 / 37373 (AAD)
Description
Includes
1 inlet plug and 2 seals, 1 outlet plug and 2 seals and 1 battery connector cover
- Plug and seal for inlet and outlet openings of TR-600 series motor/blower
- Cover for electrical connections of TR-630 or TR-632 battery
- Motor/blower plugs allow motor/blower to be submerged, rinsed under running water, or machine washed
- Plug seals to be replaced after 30 uses or whenever found to be damaged or worn
- 3M Catalog# TR-653
Applications:
Recommended Industry: Healthcare, Pharmaceutical, Pesticide Application, Heavy Industrial, Animal processing, Laboratories, Nuclear energy, Manufacturing, Cosmetics, Pharmaceutical research and production, Recycling, General Manufacturing, Dry Foods
Certifications
NIOSH Approved
Specifications
Specifications
Disposable | Reusable |
Product Type | Cleaning and Storage Plug |
For Use With (Equipment) | TR-600/TR-800 PAPR System |
Includes | 1 inlet plug and 2 seals, 1 outlet plug and 2 seals, and 1 battery connector cover |
Model | TR-653 |
Quantity | 1 Ea. |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
3M™ Versaflo™ TR-600/800 PAPR Accessory, Cleaning and Storage Kit TR-653 / 37373 (AAD) >
Spot an opportunity for improvement?Share a Content Correction