missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ 4 - 20 mA Current Loop Interface

4-20mA current loop interface Humidity controller 0-100% RH, actual value

1815.00 DKK

Specifications

Description Requires option K7 or K6
View More Specs

Products 4
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17129974
View Documents
Memmert™
ICO150V7
ICO150
1815.00 DKK
Each
Estimated Shipment: 07-07-2025
to see stock.
Add to basket
 
17160005
View Documents
Memmert™
ICO240V7
ICO240
1815.00 DKK
Each
Estimated Shipment: 07-07-2025
to see stock.
Add to basket
 
17190035
View Documents
Memmert™
ICO50V7
ICO50
1815.00 DKK
Each
Estimated Shipment: 07-07-2025
to see stock.
Add to basket
 
17199934
View Documents
Memmert™
ICO105V7
ICO105
1815.00 DKK
Each
Estimated Shipment: 07-07-2025
to see stock.
Add to basket
 
Description

Description

  • Requires option K7 or K6
  • Humidity controller
  • Actual value (0 to 100 % rh = 4 - 20 mA)
Specifications

Specifications

Requires option K7 or K6
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Memmert™ 4 - 20 mA Current Loop Interface

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.