missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ 4 - 20 mA Current Loop Interface
4-20mA current loop interface Temperature controller, actual value
1810.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
17119974
|
Memmert™
ICO150V3 |
ICO150 |
1810.00 DKK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
17150005
|
Memmert™
ICO240V3 |
ICO240 |
1810.00 DKK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
17180035
|
Memmert™
ICO50V3 |
ICO50 |
1810.00 DKK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
17189934
|
Memmert™
ICO105V3 |
ICO105 |
1810.00 DKK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
Description
- Temperature controller
- Actual value; 0 to +70°C = 4 - 20 mA
Specifications
Temperature Controller |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ 4 - 20 mA Current Loop Interface