Learn More
Memmert™ 4-20 mA Current Loop Interface for UF260
Temperature Output for external temperature monitoring.
Brand: Memmert™ UF260V6
This item is not returnable.
View return policy
Description
- Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
UF260 Oven | |
Current Loop Interface | |
4 to 20 mA |
Product Content Correction
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF260 > For Use With: UF260
Spot an opportunity for improvement?Share a Content Correction