Learn More
Memmert™ 4-20 mA Current Loop Interface for UF55
Temperature Output for external temperature monitoring.
Brand: Memmert™ UF55V6
3267.90 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
UF55 Oven | |
Current Loop Interface | |
4 to 20 mA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF55 > For Use With: UF55
Spot an opportunity for improvement?Share a Content Correction