Learn More
Memmert™ 4-20 mA Current Loop Interface for UF55, Actual Value
Temperature Output for external temperature monitoring.
Brand:  Memmert™ UF55V3
Product Code. 17569525
  • 1690.00 DKK / Each
Only null left
Estimated Shipment: 18-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Temperature controller actual value
Specifications
Specifications
UF55 Oven
Current Loop Interface
4 to 20 mA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF55, Actual Value > For Use With: UF55