Learn More
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For Vo29
Memmert™ Temperature Output for external temperature monitoring.
Brand:  Memmert™ VO29V6
Product Code. 16354557
  • 3355.00 DKK / Each
Estimated Shipment: 19-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • 4 to 20 mA current loop interface
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring
Specifications
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Current Loop Interface
4 to 20 mA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For Vo29 >