missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-2,3,5,6-tetrafluorobenzoic acid, 97+% Product Code.: 15495767

4-Bromo-2,3,5,6-tetrafluorobenzoic acid, 97+%

Product Code. 15495767
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15495767

Brand: Thermo Scientific Alfa Aesar H32982.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4707-24-8 | C7HBrF4O2 | 272.981 g/mol

4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4707-24-8
Molecular Formula C7HBrF4O2
Molecular Weight (g/mol) 272.981
MDL Number MFCD00075244
InChI Key MTZLICDXLGQWJV-UHFFFAOYSA-N
Synonym 4-bromo-2,3,5,6-tetrafluoro-benzoic acid, acmc-1arsf, 4-bromotetrafluorobenzoic acid, 4-bromotetrafluorobenzene carboxylic acid, benzoicacid, 4-bromo-2,3,5,6-tetrafluoro, benzoic acid, 4-bromo-2,3,5,6-tetrafluoro, 4-bromo-2,3,5,6-tetrafluorobenzoic acid, 4-bromanyl-2,3,5,6-tetrakis fluoranyl benzoic acid
PubChem CID 298173
IUPAC Name 4-bromo-2,3,5,6-tetrafluorobenzoic acid
SMILES C1(=C(C(=C(C(=C1F)F)Br)F)F)C(=O)O

Specifications

Melting Point 142°C to 145°C
Odor Odorless
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 272.98
Percent Purity ≥97%
Chemical Name or Material 4-Bromo-2,3,5,6-tetrafluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-2,3,5,6-tetrafluorobenzoic acid, 97+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.