missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 4-Chloro-D-phenylalanine, 95% Product Code.: 15438467

Thermo Scientific Chemicals 4-Chloro-D-phenylalanine, 95%

Product Code. 15438467
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15438467

Brand: Thermo Scientific Chemicals H51982.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is applied in the synthesis and placental binding potencies of photosensitive analogues of luteinizing hormone releasing hormone (lhrh) with agonistic and antagonistic structures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the synthesis and placental binding potencies of photosensitive analogues of luteinizing hormone releasing hormone (lhrh) with agonistic and antagonistic structures.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14091-08-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00079675
InChI Key NIGWMJHCCYYCSF-UHFFFAOYNA-N
Synonym 4-chloro-d-phenylalanine, d-4-chlorophenylalanine, r-2-amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro, 2r-2-amino-3-4-chlorophenyl propanoic acid, h-d-phe 4-cl-oh, fenclonine, r, d-p-chlorophenylalanine, unii-rss1jq7mes, rss1jq7mes
PubChem CID 667429
IUPAC Name (2R)-2-amino-3-(4-chlorophenyl)propanoic acid
SMILES NC(CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point 260°C
UN Number UN2811
Beilstein 2416151
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 199.64
Percent Purity 95%
Chemical Name or Material 4-Chloro-D-phenylalanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ01B007 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51982.MD,H51982.06,H51982.03
Lot NumberP19E029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51982.MD,H51982.06,H51982.03
Lot NumberN16H021 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51982.MD,H51982.06,H51982.03
Lot NumberT09F034 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51982.MD,H51982.06,H51982.03
Lot NumberK19W027 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH51982.MD,H51982.06,H51982.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 4-Chloro-D-phenylalanine, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.