missing translation for 'onlineSavingsMsg'
Learn More
4-Ethoxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™ Product Code.: 15426957

4-Ethoxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 15426957
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15426957

Brand: Thermo Scientific Alfa Aesar H34048.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 252901-50-1
Molecular Formula C10H9F3O3
Molecular Weight (g/mol) 234.17
MDL Number MFCD13196578
InChI Key KWEQYUCUGSDCHR-UHFFFAOYSA-N
Synonym 4-ethoxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-ethoxy-3-trifluoromethyl
PubChem CID 23508153
IUPAC Name 4-ethoxy-3-(trifluoromethyl)benzoic acid
SMILES CCOC1=C(C=C(C=C1)C(O)=O)C(F)(F)F

Specifications

Quantity 1 g
Formula Weight 234.17
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-3-(trifluoromethyl)benzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10173722 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Ethoxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.