Learn More
4-Ethyloctanoic acid, 97%, Thermo Scientific Chemicals
Description
Five sulfur-containing flavor compounds were synthesized by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. 4-ethyloctanoic acid is identified from flue-cured Virginia tobacco. Mainly used as daily flavor,food flavor,industrial flavor, nutrient, stabilizers, surfactants and emulsifiers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Five sulfur-containing flavor compounds were synthesized by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. 4-ethyloctanoic acid is identified from flue-cured Virginia tobacco. Mainly used as daily flavor,food flavor,industrial flavor, nutrient, stabilizers, surfactants and emulsifiers.
Solubility
Slightly soluble in water(0.13 mg/mL). Soluble in hexane
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Density | 0.908 |
Boiling Point | 142°C to 143°C (10 mmHg) |
Flash Point | >100°C (212°F) |
Refractive Index | 1.439 |
Quantity | 10 g |
Solubility Information | Slightly soluble in water(0.13mg/mL). Soluble in hexane |
Formula Weight | 172.27 |
Percent Purity | 97% |
Chemical Name or Material | 4-Ethyloctanoic acid |
Safety and Handling
- 4-Ethyloctanoic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 4-Ethyloctanoic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.