missing translation for 'onlineSavingsMsg'
Learn More
4-Fluoro-2,3-dimethylaniline, 97% Product Code.: 15404157

4-Fluoro-2,3-dimethylaniline, 97%

Product Code. 15404157
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 15404157

Brand: Thermo Scientific Alfa Aesar H31560.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 1737-68-4 | C8H10FN | 139.173 g/mol

4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2,3-dimethylaniline is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1737-68-4
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD04972870
InChI Key LKMJLYCMLAPNDB-UHFFFAOYSA-N
Synonym 3-amino-6-fluoro-1,2-dimethylbenzene, 4-fluoro-2,3-dimethyl aniline, 2,3-dimethyl-4-fluoroaniline, 4-fluoro-o-xylidine, benzenamine, 4-fluoro-2,3-dimethyl, 4-fluoro-2,3-dimethyl-phenylamine, p-fluordimethylanilin, intermediates-zcf02293, 4-fluoranyl-2,3-dimethyl-aniline, 4-fluoro-2,3-dimethyl-benzenamine
PubChem CID 17770833
IUPAC Name 4-fluoro-2,3-dimethylaniline
SMILES CC1=C(C=CC(=C1C)F)N

Specifications

Melting Point 42°C to 46°C
Quantity 250 mg
UN Number UN2811
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2,3-dimethylaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Product Code
Lot Number10247417 Certificate TypeCertificate of Analysis Date20/07/2023 Product CodeH31560.MD,H31560.03,H31560
Lot Number10157348 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH31560.MD,H31560.03
Lot Number10199700 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH31560.MD,H31560.03
Lot Number10223936 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH31560.MD,H31560.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluoro-2,3-dimethylaniline, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.