missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenylacetic acid, 98+%, Thermo Scientific Chemicals

543.62 DKK - 1885.37 DKK

Chemical Identifiers

CAS 4476-28-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00041035
InChI Key RERBQXVRXYCGLT-UHFFFAOYSA-N
Synonym 4-isopropylphenylacetic acid, 2-4-isopropylphenyl acetic acid, 4-isopropylphenyl acetic acid, benzeneacetic acid, 4-1-methylethyl, 2-4-propan-2-yl phenyl acetic acid, p-isopropyl phenyl acetic acid, 4-isopropyl-phenyl-acetic acid, 4-propan-2-yl phenyl acetic acid, 2-4-methylethyl phenyl acetic acid, 4-isopropylphenylaceticacid
PubChem CID 78230
IUPAC Name 2-(4-propan-2-ylphenyl)acetic acid
SMILES CC(C)C1=CC=C(C=C1)CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361817
View Documents
Thermo Scientific Alfa Aesar
L05071.03
1 g
543.62 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
11371817
View Documents
Thermo Scientific Alfa Aesar
L05071.06
5 g
1885.37 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4476-28-2
178.231
RERBQXVRXYCGLT-UHFFFAOYSA-N
78230
CC(C)C1=CC=C(C=C1)CC(=O)O
C11H14O2
MFCD00041035
4-isopropylphenylacetic acid, 2-4-isopropylphenyl acetic acid, 4-isopropylphenyl acetic acid, benzeneacetic acid, 4-1-methylethyl, 2-4-propan-2-yl phenyl acetic acid, p-isopropyl phenyl acetic acid, 4-isopropyl-phenyl-acetic acid, 4-propan-2-yl phenyl acetic acid, 2-4-methylethyl phenyl acetic acid, 4-isopropylphenylaceticacid
2-(4-propan-2-ylphenyl)acetic acid

Specifications

4476-28-2
C11H14O2
2328756
RERBQXVRXYCGLT-UHFFFAOYSA-N
2-(4-propan-2-ylphenyl)acetic acid
78230
≥98%
51°C to 52°C
MFCD00041035
4-isopropylphenylacetic acid, 2-4-isopropylphenyl acetic acid, 4-isopropylphenyl acetic acid, benzeneacetic acid, 4-1-methylethyl, 2-4-propan-2-yl phenyl acetic acid, p-isopropyl phenyl acetic acid, 4-isopropyl-phenyl-acetic acid, 4-propan-2-yl phenyl acetic acid, 2-4-methylethyl phenyl acetic acid, 4-isopropylphenylaceticacid
CC(C)C1=CC=C(C=C1)CC(=O)O
178.231
178.23
4-Isopropylphenylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 224-755-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.