missing translation for 'onlineSavingsMsg'
Learn More
4-(Methoxycarbonylamino)benzeneboronic acid, 97%, Thermo Scientific Chemicals Product Code.: 15489797

4-(Methoxycarbonylamino)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15489797
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15489797

Brand: Thermo Scientific Alfa Aesar H53027.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850567-96-3
Molecular Formula C8H10BNO4
Molecular Weight (g/mol) 194.981
MDL Number MFCD03788425
InChI Key ROAKVYQVTMOEGR-UHFFFAOYSA-N
Synonym 4-methoxycarbonylamino phenylboronic acid, 4-methoxycarbonyl amino phenyl boronic acid, 4-methoxycarbonylamino benzeneboronic acid, 4-methoxycarbonyl amino phenylboronic acid, 4-methoxycarbonylamino phenyl boronic acid, acmc-209q0y, 4-methoxycarbonylamino-phenyl boronic acid, 4-methoxycarbonylamino benzene boronic acid, 4-methoxycarbonyl amino phenyl boronicacid
PubChem CID 2773494
IUPAC Name [4-(methoxycarbonylamino)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)NC(=O)OC)(O)O

Specifications

Melting Point 234°C to 236°C
Quantity 1 g
Formula Weight 194.98
Percent Purity 97%
Chemical Name or Material 4-(Methoxycarbonylamino)benzeneboronic Acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW06J007 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberH53027.MD,H53027.03
Lot NumberZ13D008 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53027.MD,H53027.03
Lot NumberF10Z018 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53027.MD,H53027.03
Lot NumberQ14D003 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53027.MD,H53027.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Methoxycarbonylamino)benzeneboronic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.