Learn More
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11487007
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals
Product Code. 11487007
100 g
Click to view available options
Quantity:
5 g
100 g
500 g
Product Code. 11487007
Brand: Thermo Scientific Alfa Aesar A14399.22
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104-01-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00004345
InChI Key NRPFNQUDKRYCNX-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetic acid, homoanisic acid, 2-4-methoxyphenyl acetic acid, 4-methoxybenzeneacetic acid, benzeneacetic acid, 4-methoxy, 4-methoxyphenyl acetic acid, p-methoxyphenylacetic acid, 2-p-anisyl acetic acid, p-methoxyphenyl acetic acidShow MoreShow Less
PubChem CID 7690
ChEBI CHEBI:55501
IUPAC Name 2-(4-methoxyphenyl)acetic acid
SMILES COC1=CC=C(C=C1)CC(=O)O
Specifications
Melting Point 84°C to 88°C
Boiling Point 138°C to 140°C (3 mmHg)
Quantity 100 g
Beilstein 1101737
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 4-Methoxyphenylacetic acid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449367 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14399.36,A14399.06,A14399.22
Lot NumberA0448223 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberA14399.36,A14399.06,A14399.22
Lot NumberA0439162 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberA14399.36,A14399.06,A14399.22
Lot Number10239717 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14399.36,A14399.06,A14399.22
Lot Number10237900 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14399.36,A14399.06,A14399.22
compliance-icons
Product Identifier
  • 4-Methoxyphenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Methoxyphenylacetic acid
RUO – Research Use Only
Product Content Correction
Product Title
4-Methoxyphenylacetic acid, 98+%, Thermo Scientific Chemicals >