missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Morpholinobenzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Synonym | 4-morpholinobenzonitrile, 4-4-cyanophenyl morpholine, 4-morpholin-4-yl benzonitrile, 4-4-morpholinyl benzonitrile, 4-morpholin-4-yl-benzonitrile, benzonitrile, 4-4-morpholinyl, benzonitrile,4-4-morpholinyl, 4-morpholin-4-ylbenzenecarbonitrile, pubchem18708, acmc-1bqst |
Formula Weight | 188.23 |
Percent Purity | 97% |
Chemical Name or Material | 4-Morpholinobenzonitrile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Morpholinobenzonitrile, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction