Learn More
4-Piperidinemethanol, 98%
CAS: 6457-49-4 | C6H13NO | 115.17 g/mol
652.00 DKK - 5747.00 DKK
Chemical Identifiers
CAS | 6457-49-4 |
---|---|
Molecular Formula | C6H13NO |
Molecular Weight (g/mol) | 115.17 |
MDL Number | MFCD00174228 |
InChI Key | XBXHCBLBYQEYTI-UHFFFAOYSA-N |
Synonym | 4-piperidinemethanol, 4-hydroxymethyl piperidine, 4-piperidylmethanol, 4-hydroxymethylpiperidine, piperidin-4-yl methanol, piperidin-4-yl-methanol, piperidine-4-methanol, 4-piperidylmethan-1-ol, 4-piperidine methanol, 4-piperidyl carbinol |
PubChem CID | 420771 |
IUPAC Name | piperidin-4-ylmethanol |
SMILES | C1CNCCC1CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10220013
|
Thermo Scientific Acros
380630050 |
5 g | Glass bottle |
652.00 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10564673
|
Thermo Scientific Acros
380630250 |
25 g | Glass bottle |
2200.00 DKK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10217033
|
Thermo Scientific Acros
380631000 |
100 g | Glass bottle | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6457-49-4 | |
115.17 | |
XBXHCBLBYQEYTI-UHFFFAOYSA-N | |
420771 | |
C1CNCCC1CO |
C6H13NO | |
MFCD00174228 | |
4-piperidinemethanol, 4-hydroxymethyl piperidine, 4-piperidylmethanol, 4-hydroxymethylpiperidine, piperidin-4-yl methanol, piperidin-4-yl-methanol, piperidine-4-methanol, 4-piperidylmethan-1-ol, 4-piperidine methanol, 4-piperidyl carbinol | |
piperidin-4-ylmethanol |
Specifications
6457-49-4 | |
100.0 | |
White | |
>110°C | |
97.5% min. (GC) | |
MFCD00174228 | |
XBXHCBLBYQEYTI-UHFFFAOYSA-N | |
piperidin-4-ylmethanol | |
420771 | |
98% | |
4-Piperidinemethanol |
97.5 | |
57.0°C to 61.0°C | |
118.0°C to 120.0°C (10.0 mmHg) | |
Authentic | |
C6H13NO | |
4-piperidinemethanol, 4-hydroxymethyl piperidine, 4-piperidylmethanol, 4-hydroxymethylpiperidine, piperidin-4-yl methanol, piperidin-4-yl-methanol, piperidine-4-methanol, 4-piperidylmethan-1-ol, 4-piperidine methanol, 4-piperidyl carbinol | |
C1CNCCC1CO | |
115.17 | |
115.17 | |
Crystalline Powder, Chunks or Solid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
Danger
- 4-piperidylmethanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.