missing translation for 'onlineSavingsMsg'
Learn More

4-PPBP maleate, Tocris Bioscience™

Product Code. 18004392
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Specific σ ligand, NMDA antagonist

σ ligand and selective non-competitive antagonist at recombinant NR1a/2B NMDA receptors expressed in Xenopus oocytes.

Chemical Identifiers

CAS 201216-39-9
Molecular Formula C25H31NO4
Molecular Weight (g/mol) 409.526
InChI Key OASPNIMFGJVLES-WLHGVMLRSA-N
Synonym 4-ppbp maleate, 4-phenyl-1-4-phenylbutyl piperidine maleate salt
PubChem CID 71300190
IUPAC Name (E)-but-2-enedioic acid;4-phenyl-1-(4-phenylbutyl)piperidine
SMILES C1CN(CCC1C2=CC=CC=C2)CCCCC3=CC=CC=C3.C(=CC(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 409.52
Percent Purity >99%
Chemical Name or Material 4-PPBP maleate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'