Learn More
BRAND™ BRANDplates™ Microplate Insert System With Inlet Opening System
Six well plates filled with six insert strips
Brand:  BRAND™ 782873
 View more versions of this product
Product Code. 15372265
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Sterile
  • With inlet opening system
  • cellGrade™ plus surface
  • Membrane material: PC or PET material
  • Multiple membrane pore sizes for different applications: 0.4μm, 1.0μm, 3.0μm, 8.0μm
  • 5 plates/Pk. with lids and 30 insert strips
Applications:
in vitro preparation of tissue (3D tissue culture)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
Microplate Insert System w/Inlet Opening System
8 μm
PET
cellGrade plus
6
5/Pk.