missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

173.00 DKK - 2045.00 DKK

Chemical Identifiers

CAS 215434-25-6
Molecular Formula C8H6ClNO2S3
Molecular Weight (g/mol) 279.771
InChI Key YIKNRCVJBOLZFL-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonyl chloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-2-methyl-4-thiazolyl, 5-2-methylthiazol-4-yl thiophene-2-sulphonyl chloride, 5-2-methyl-1,3-thiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulfonylchloride, 5-2-methylthiazol-4-yl thiophene-2-sulphonylchloride, 5-2-methyl-thiazol-4-yl-thiophene-2-sulfonyl chloride, 5-2-methyl-1,3-thiazol-4-yl-2-thiophenesulfonyl chloride, chloro 5-2-methyl 1,3-thiazol-4-yl 2-thienyl sulfone
PubChem CID 2736979
IUPAC Name 5-(2-methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride
SMILES CC1=NC(=CS1)C2=CC=C(S2)S(=O)(=O)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10590552
SDS View Documents
Thermo Scientific Maybridge
CC18203CB
250 mg
173.00 DKK
250mg
Please to purchase this item. Need a web account? Register with us today!
10374322
SDS View Documents
Thermo Scientific Maybridge
CC18203DA
1 g
567.00 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
10727285
SDS View Documents
Thermo Scientific Maybridge
CC18203DE
5 g
2045.00 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(2-Methyl-1,3-thiazol-4-yl)thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.