Learn More
5-(4-Hydroxyphenyl)pentanoic acid, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
5-(4-Hydroxyphenyl)pentanoic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Specifications
Specifications
Melting Point | 116°C to 120°C |
Molecular Formula | C11H14O3 |
MDL Number | MFCD04039777 |
Quantity | 1 g |
Synonym | 5-4-hydroxyphenyl pentanoic acid, 5-4-hydroxyphenyl valeric acid, pubchem16852, acmc-20am8d, 5-4-hydroxyphenyl pentanoicacid, benzenepentanoic acid, 4-hydroxy |
Solubility Information | Soluble in water. |
InChI Key | YSSJQFONKASLKM-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1CCCCC(=O)O)O |
IUPAC Name | 5-(4-hydroxyphenyl)pentanoic acid |
Molecular Weight (g/mol) | 194.23 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
10121966 | Certificate of Analysis | 07/12/2021 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.