missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 5,7-Dichlorokynurenic acid sodium salt
Sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286). Potent NMDA antagonist, acts at glycine site
1130.00 DKK - 4170.00 DKK
Specifications
Purity | >98% |
---|---|
Product Type | 5,7-Dichlorokynurenic acid sodium salt |
Molecular Weight (g/mol) | 280.04 |
Inhibitors | 5,7-Dichlorokynurenic acid sodium salt |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18024082
|
Tocris Bioscience™
3698/10 |
10 mg |
1130.00 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18085782
|
Tocris Bioscience™
3698/50 |
50 mg |
4170.00 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286), a potent antagonist at the glycine site of the NMDA receptor (Ki = 79 nM vs. [3H]-glycine).Specifications
>98% | |
5,7-Dichlorokynurenic acid sodium salt | |
280.04 | |
5,7-Dichlorokynurenic acid sodium salt |
For Research Use Only