missing translation for 'onlineSavingsMsg'
Learn More
5-Chloro-2-iodoaniline, 98%, Thermo Scientific Chemicals Product Code.: 11399919

5-Chloro-2-iodoaniline, 98%, Thermo Scientific Chemicals

Product Code. 11399919
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 11399919

Brand: Thermo Scientific Alfa Aesar B25709.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6828-35-9
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.467
MDL Number MFCD06738960
InChI Key FEOMAFDDLHSVMO-UHFFFAOYSA-N
Synonym benzenamine, 5-chloro-2-iodo, 5-chloro-2-iodo-phenylamine, 5-chloro-2-iodo-benzenamine, 5-chloro-2-iodophenylamine, 2-amino-4-chloro-iodobenzene, pubchem5286, 2-iodo-5-chloroaniline, 5-chloro-2-iodoanilline, 5-chloro-2-iodo-aniline, acmc-209o2s
PubChem CID 639697
IUPAC Name 5-chloro-2-iodoaniline
SMILES C1=CC(=C(C=C1Cl)N)I

Specifications

Melting Point 40°C to 42°C
Quantity 5 g
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 253.47
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-iodoaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number10144401 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB25709.03,B25709.06
Lot Number10103313 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeB25709.03,B25709.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Chloro-2-iodoaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.