missing translation for 'onlineSavingsMsg'
Learn More
5-Decyn-1-ol, 97%, Thermo Scientific Chemicals Product Code.: 15480168

5-Decyn-1-ol, 97%, Thermo Scientific Chemicals

Product Code. 15480168
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15480168

Brand: Thermo Scientific Alfa Aesar H53400.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Decyn-1-ol is used in the synthesis of multi-substituted cyclopropanes. It involved in manufacture of fine chemicals, having applications in electroplating and perfumery industry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68274-97-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00464353
InChI Key RNTNGQCWAKHVKO-UHFFFAOYSA-N
Synonym 5-decyn-1-ol, 5-decyn-1-ol,
PubChem CID 109957
IUPAC Name dec-5-yn-1-ol
SMILES CCCCC#CCCCCO

Specifications

Density 0.861
Boiling Point 238°C
Linear Formula CH3(CH2)3C≡C(CH2)4OH
Refractive Index 1.461
Quantity 25 g
Formula Weight 154.25
Percent Purity 97%
Chemical Name or Material 5-Decyn-1-ol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM23K027 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog NumberH53400.14,H53400.06
Lot NumberM14I023 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberH53400.14,H53400.06
Lot NumberE25X011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH53400.14,H53400.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Decyn-1-ol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.