missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrophenol, 98%, Thermo Scientific Chemicals

Product Code. 11414274
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-36-6
Molecular Formula C6H4FNO3
Molecular Weight (g/mol) 157.1
MDL Number MFCD00007107
InChI Key QQURWFRNETXFTN-UHFFFAOYSA-N
Synonym 2-nitro-5-fluorophenol, 3-fluoro-6-nitrophenol, phenol, 5-fluoro-2-nitro, 4-fluoro-2-hydroxynitrobenzene, 5-fluoro-2-nitro-phenol, pubchem1490, pubchem2371, acmc-209jzc, 3-fluoro 6-nitrophenol, 5-flouro-2-nitrophenol
PubChem CID 9937
IUPAC Name 5-fluoro-2-nitrophenol
SMILES C1=CC(=C(C=C1F)O)[N+](=O)[O-]

Specifications

Melting Point 30°C to 36°C
Boiling Point 260°C
Flash Point 91°C (195°F)
Quantity 10 g
Beilstein 1870311
Formula Weight 157.1
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-nitrophenol
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'