missing translation for 'onlineSavingsMsg'
Learn More

5-(Tri-n-butylstannyl)pyrimidine, 96%

Product Code. 15428097
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 144173-85-3 | C16H30N2Sn | 369.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144173-85-3
Molecular Formula C16H30N2Sn
Molecular Weight (g/mol) 369.14
MDL Number MFCD01319056
InChI Key QRDQHTJNKPXXRQ-UHFFFAOYSA-N
Synonym 5-tributylstannyl pyrimidine, pyrimidine, 5-tributylstannyl, tributyl pyrimidin-5-yl stannane, 5-tri-n-butylstannyl pyrimidine, acmc-20dn5a, 5-tributylstannylpyrimidine, 5-tributylstannanyl-pyrimidine, tri-n-butyl 5-pyrimidinyl tin, 5-tri-butylstannyl pyrimidine, tributyl 5-pyrimidinyl stannane
PubChem CID 2763249
IUPAC Name tributyl(pyrimidin-5-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=CN=C1

Specifications

Quantity 1 g
UN Number UN2788
Formula Weight 369.13
Percent Purity 96%
Chemical Name or Material 5-(Tri-n-butylstannyl)pyrimidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(Tri-n-butylstannyl)pyrimidine, 96% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.