missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium Standard: Pd @ 10000 μg/mL in 20% HCl, VHG™
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Description
Palladium Standard: Pd @ 10000 ÎĽg/mL in 20% HCl
Specifications
Specifications
Chemical Name or Material | Palladium |
CAS | 7440-05-3 |
Methods | ICP/ICP-MS |
Analyte or Component Names | Palladium |
Shipping Condition | Room Temperature |
Molecular Formula | Pd |
Concentration | 10000 ÎĽg/mL |
Certifications/Compliance | ISO 17034 |
Synonym | Palladium on carbon, Palladium Black, Paladio, Palladium/carbon, Palladium, IG 0218A, MPP 030, MPP 050, MPP 080, P 50, P 50 (metal), Palladex 600, Palladium (nanoparticles), Palladium (powder), Palladium element, SFP 1001P, TCP 211 |
InChI Formula | InChI=1S/Pd |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Palladium Standard: Pd @ 10000 μg/mL in 20% HCl, VHG™ >
Spot an opportunity for improvement?Share a Content Correction