Learn More
Cellpath Stainless-steel Reusable Base Molds
Ideal for prostrate and breast cores and can be used with both grid and slatted processing cassettes for both horizontal and vertical sectioning. Cellpath Stainless-steel Reusable Base Moulds are manufactured from high quality stainless steel that put an end to extensive trimming of wax.
Brand:  Cellpath GBB-3014-05A
 View more versions of this product
Product Code. 30226275
  • 327.54 DKK / Pack of 5
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Made of reusable Stainless steel
  • Stackable to reduce storage area
  • Internal rounded corners enable easy removal of the wax block
  • Features extended edges prevent tipping and provide grip area for ease of handling
  • Available in wide range of sizes
  • GBB-0714-05A suitable for use with needle and punch biopsies
Specifications
Specifications
Stainless Steel
30 x 24 x 9 mm
5/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cellpath Stainless-steel Reusable Base Molds > Size: 30 x 24 x 9mm