Learn More
BTX™ 6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo
6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo are miniature needles designed to provide highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse In Vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.
Brand:  BTX™ 470080
 View more versions of this product
Product Code. 15437380
  • 2810.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
  • Uniform, reliable electric fields
  • Miniature needles minimize tissue trauma
  • Disposable and sterile
  • Medical-grade plastic and surgical steel construction
  • Safety-assured design
  • Multiple configurations, sizes to choose from
Specifications
Specifications
Gene and Vaccine Delivery
Stainless Steel
Electroporation Needle Array
0.03 in.
Straight/Needle Array
0.98 in.
1 Ea.
Trocar Point
Electrode Connector Cable
AgilePulse™ in Vivo Gene Delivery Electroporation System
BTX Electroporation
6-needle Array, 6 mm gap, 25 mm length
0.7 mm
6-needle Array
25 mm
6 mm
Sterile
Square Wave
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Intended for Research use only. Not for use in diagnostic or clinical procedures.