Learn More
6-Phenyl-2-thiouracil-13C,15N2, Honeywell Fluka™ Product Code.: 15949761
6-Phenyl-2-thiouracil-13C,15N2, Honeywell Fluka™
Product Code. 15949761 Shop All Honeywell Fluka Products
10 mg, 10mg
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 15949761
Brand: Honeywell Fluka R150110MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Analytical standard, reference material
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY
Specifications
Packaging Glass Bottle
Physical Form Solid
Industry Type Steroids and Hormones
Quantity 10 mg
Grade Analytical Standard
Product Content Correction
Product Title
6-Phenyl-2-thiouracil-13C,15N2, Honeywell Fluka™ >