Learn More
6-(Tetrahydropyran-4-yloxy)nicotinic acid, 97%, Thermo Scientific™
755.00 DKK - 950.00 DKK
Chemische Identifikatoren
CAS | 886851-55-4 |
---|---|
Molecular Formula | C11H13NO4 |
Molecular Weight (g/mol) | 223.228 |
MDL Number | MFCD09258833 |
InChI Key | OLOSQIMMFCSJHD-UHFFFAOYSA-N |
Synonym | 6-oxan-4-yloxy pyridine-3-carboxylic acid, 6-tetrahydropyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yl oxy nicotinic acid, 2-tetrahydro-2h-pyran-4-yloxy pyridine-5-carboxylic acid, 3-pyridinecarboxylicacid, 6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yl oxy pyridine-3-carboxylic acid, 4-5-carboxypyridin-2-yl oxy tetrahydro-2h-pyran |
PubChem CID | 24229576 |
IUPAC Name | 6-(oxan-4-yloxy)pyridine-3-carboxylic acid |
SMILES | C1COCCC1OC2=NC=C(C=C2)C(=O)O |
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Packaging | Preis | Menge & Verfügbarkeit | ||||
11306815
|
Thermo Scientific Maybridge
CC58301CB |
250 mg | Amber Glass Bottle |
755.00 DKK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10328063
|
Thermo Scientific Maybridge
CC58301DA |
1 g | Amber Glass Bottle |
950.00 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Chemische Identifikatoren
886851-55-4 | |
223.228 | |
OLOSQIMMFCSJHD-UHFFFAOYSA-N | |
24229576 | |
C1COCCC1OC2=NC=C(C=C2)C(=O)O |
C11H13NO4 | |
MFCD09258833 | |
6-oxan-4-yloxy pyridine-3-carboxylic acid, 6-tetrahydropyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yl oxy nicotinic acid, 2-tetrahydro-2h-pyran-4-yloxy pyridine-5-carboxylic acid, 3-pyridinecarboxylicacid, 6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yl oxy pyridine-3-carboxylic acid, 4-5-carboxypyridin-2-yl oxy tetrahydro-2h-pyran | |
6-(oxan-4-yloxy)pyridine-3-carboxylic acid |
Spezifikation
886851-55-4 | |
100.0 | |
C11H13NO4 | |
6-oxan-4-yloxy pyridine-3-carboxylic acid, 6-tetrahydropyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yloxy nicotinic acid, 6-tetrahydro-2h-pyran-4-yl oxy nicotinic acid, 2-tetrahydro-2h-pyran-4-yloxy pyridine-5-carboxylic acid, 3-pyridinecarboxylicacid, 6-tetrahydro-2h-pyran-4-yl oxy, 6-oxan-4-yl oxy pyridine-3-carboxylic acid, 4-5-carboxypyridin-2-yl oxy tetrahydro-2h-pyran | |
C1COCCC1OC2=NC=C(C=C2)C(=O)O | |
223.228 | |
223.23 | |
6-(Tetrahydropyran-4-yloxy)nicotinic acid |
96.5 | |
97% | |
MFCD09258833 | |
OLOSQIMMFCSJHD-UHFFFAOYSA-N | |
6-(oxan-4-yloxy)pyridine-3-carboxylic acid | |
24229576 | |
97% |
Sicherheit und Handhabung
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning