missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals

Product Code. 15497508
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

7-Bromo-3,4-dihydro-2H-1,4-benzothiazine used in many areas of medicine. They are used in industries as dyes and as antioxidants of rubber and natural elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine used in many areas of medicine. They are used in industries as dyes and as antioxidants of rubber and natural elastomers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193414-60-7
Molecular Formula C8H8BrNS
Molecular Weight (g/mol) 230.12
MDL Number MFCD11847754
InChI Key QWWYGAWGUCQKDL-UHFFFAOYSA-N
Synonym 7-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine
PubChem CID 83823811
IUPAC Name 7-bromo-3,4-dihydro-2H-1,4-benzothiazine
SMILES BrC1=CC2=C(NCCS2)C=C1

Specifications

Quantity 250 mg
Formula Weight 230.12
Percent Purity 97%
Chemical Name or Material 7-Bromo-3,4-dihydro-2H-1,4-benzothiazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'