Learn More
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid
GREENER_CHOICE
Hold eight-one 1.5 or 2.0mL cryogenic storage vials with this stackable cryogenic storage rack with lid.
Brand:  Thermo Scientific™ 8820
2366.81 DKK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
 View more versions of this product
Product Code. 10781592
  • 2665.00 DKK / Pack of 20
Only null left
Estimated Shipment: 21-07-2025
Description
Hold eighty-one 1.5 or 2.0mL cryogenic storage vials with the Thermo Scientific™ 81-Well Cryogenic Storage Rack with Lid. Available in several colors with transparent tinted lid that protects samples from dust and debris and permits stacking.
  • Molded with alphanumeric grid for easy tube identification
  • Lid protects samples and allows racks to be stacked
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fluorescent Orange
81 (1.5 mL or 2.0 mL Tubes)
Polypropylene
81-Well Cryogenic Storage Racks with Lid
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ 81-Well Cryogenic Storage Racks with Lid > Cryogenic Rack, Fluorescent Orange