Learn More
Fisherbrand™ 8mm PP Screw Cap, 8-425
Polypropylene caps are chemically inert and suitable for most chromatography applications.
Brand: Fisherbrand™ 08 08 0420
386.33 DKK valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
- 8mm
- PP
- Screw Cap
- 8-425 thread
Specifications
Black | |
Polypropylene | |
Closed Top Screw Cap |
8 mm | |
100 | |
ND8 |
Accessories and Services
-
Fisherbrand™ Septum for 8-425 screw thread cap X1000 8mm Septa, Silicone cream/PTFE red... Product Code: 11830772
1805.00 DKK / Pack of 1000 -
Fisherbrand™ Septum for 8-425 screw thread cap X1000 8mm Septa, Silicone dark blue/PTFE... Product Code: 11840772
2215.00 DKK / Pack of 1000 -
Fisherbrand™ Septum for 8-425 screw thread cap X1000 8mm Septa, Silicone transparent bl... Product Code: 11850772 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
-
Fisherbrand™ Septum for 8-425 screw thread cap X1000 8mm Septa, Silicone white/PTFE red... Product Code: 11872663
1805.00 DKK / Pack of 1000
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31