Description
- Brown tinted polycarbonate replacement lens
- Supravision sapphire coating
Supravision Sapphire Coating
- Inside: Scratch-resistant and chemical-resistant
- Outside: Scratch-resistant and chemical-resistant
- Non-stick properties allow for easy removal of greasy/watery dirt
Specifications
Specifications
Certifications/Compliance | EN 166:2001, EN 170:2002 |
Lens Type | Supravision Sapphire |
Lens Tint | Brown |
Disposable | Reusable |
Lens Material | Polycarbonate |
Labeling | 5-2.5 |
Product Content Correction
Product Title
Uvex™ 9160 I-Vo Replacement Lens >
Spot an opportunity for improvement?Share a Content Correction