Learn More
uvex 9178 Super Fit Safety Spectacles
Shop All Uvex Products
Click to view available options
Supravision Sapphire
Description
- Ultra-thin, wrap-around lenses with 6.5 base curvature
- A high level of protection against mechanical hazards
- Lightweight design with hinged side arms and a sporty design
- Aerodynamically optimized lenses are angled forward for comfort and ventilation
- Maximum optical clarity
Supravision Sapphire Coating
- Inside: Scratch-resistant and chemical-resistant
- Outside: Scratch-resistant and chemical-resistant
- Non-stick properties allow for easy removal of greasy/watery dirt
Specifications
Specifications
Certifications/Compliance | EN 166:2001, EN 170:2002 |
Lens Type | Aerodynamically Optimized |
Lens Coating | Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Spectacles |
Frame Color | Navy Blue, Clear |
Model | 9178 Super Fit |
Material | Polycarbonate |
Disposable | Reusable |
Lens Material | Polycarbonate |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
uvex 9178 Super Fit Safety Spectacles >
Spot an opportunity for improvement?Share a Content Correction