Description
- 96 Wells
- PP
- U-Shape Wells
- 1200μL
- Plates meet ANSI and SBS standards
- Different sealing options
- Chromatography tested
- No leeching due to high-end material
Specifications
Standard | |
Polypropylene | |
Non-sterile | |
Round Bottom (U-well) |
8 x 12 | |
96 | |
1200 μL | |
40 mm |