missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 9mm Autosampler Vial Screw Thread Caps

Prevent septa push through and over- and under-tightening. With AVCS built into these 9mm screw caps you will get an optimal, consistent seal every time.

959.00 DKK - 1975.00 DKK

Specifications

Diameter (Metric) Outer 9 mm
Material Polypropylene
View More Specs

Products 4
Product Code Brand Color Septum Price Quantity & Availability  
Product Code Brand Color Septum Price Quantity & Availability  
15266603
View Documents
Thermo Scientific™
9-SCK(R)-ST1
Red White silicone/Red PTFE This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15296603
View Documents
Thermo Scientific™
9-SCK(G)-ST1X
Green White silicone/Red PTFE This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15343425
View Documents
Thermo Scientific™
9-SCK(BLK)-BST1
Black Bonded Red PTFE/White silicone This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15353425
View Documents
Thermo Scientific™
9-SCK(GY)-BST1X
Gray Bonded Red PTFE/White silicone This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Convenience kits save time during sample preparation
  • Includes 100 vials and 100 caps with pre-assembled septa for added convenience during sample preparation
  • Reusable two compartment trays protect vials and closure while keeping matching supplies together
Specifications

Specifications

9 mm
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications