Learn More
Honeywell™ A69 Peak Series Hard Hat
Multipurpose helmet with ribbed ABS outer shell and 6-point nylon web suspension.
101.88 DKK - 181.00 DKK
Specifications
Color | White |
---|---|
Includes | Adjustable headband and suspension |
Material | Shell: ABS, Suspension: Nylon web |
Style | Ribbed, rain trough |
Product Code | Brand | Suspension Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Suspension Type | Price | Quantity & Availability | |||||
15886795
|
Honeywell™
933170 |
6-point nylon web with pin-lock adjustment | N/A | ||||||
15896795
|
Honeywell™
933180 |
6-point nylon web with ratchet adjustment |
181.00 DKK
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
Description
- Rain trough design
- Selection of pin-lock and ratchet suspensions
- Size and headband height adjustable to ensure wearer comfort
- Slots to fit face shield and earmuffs
- Easy sweatband replacement system
- Personalizing with logo printing available
- ABS shell
Specifications
White | |
Adjustable headband and suspension | |
Shell: ABS, Suspension: Nylon web | |
Ribbed, rain trough |