missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ABCB5 Antibody (Brigham and Women's patent anti-ABCB5) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28518-100ug
This item is not returnable.
View return policy
Description
ABCB5 Monoclonal antibody specifically detects ABCB5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
ABCB5 | |
Monoclonal | |
Unconjugated | |
Q2M3G0 | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
Store at -20°C in powder form. Store at -80°C once reconstituted. | |
IgG1 |
Flow Cytometry, ELISA, Functional Assay | |
Brigham and Women's patent anti-ABCB5 | |
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5 | |
ABCB5 | |
100 ÎĽg | |
ABC Transporters, Lipid and Metabolism | |
340273 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ ABCB5 Antibody (Brigham and Women's patent anti-ABCB5) - Humanized, Novus Biologicals™ > 100 μg, Unconjugated
Spot an opportunity for improvement?Share a Content Correction