missing translation for 'onlineSavingsMsg'
Learn More

AC 55541, Tocris Bioscience™

Product Code. 18045572
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective PAR2 agonist

Potent and selective protease-activated receptor 2 (PAR2) agonist (pEC50 = 6.7) that displays no activity at other PAR subtypes or at over 30 other receptors involved in nociception and inflammation. Stimulates cell proliferation, PI hydrolysis and Ca2+ mobilization in vitro (pEC50 values are 6.7, 5.9 and 6.6 respectively) and exhibits pronociceptive activity in vivo.

Chemical Identifiers

CAS 916170-19-9
Molecular Formula C25H20BrN5O3
Molecular Weight (g/mol) 518.371
InChI Key UCUHFWIFSHROPY-RWPZCVJISA-N
Synonym ac hplc , powder, 2e-2-1-3-bromophenyl ethylidene ?-benzoylamino-3,4-dihydro-4-oxo-1-phthalazineacetic acid hydrazide, e-n-2-2-1-3-bromophenyl ethylidene hydrazinyl-2-oxo-1-4-oxo-3,4-dihydrophthalazin-1-yl ethyl benzamide, n-n'-1e-1-3-bromophenyl ethylidene hydrazinecarbonyl 4-oxo-3,4-dihydrophthalazin-1-yl methyl benzamide
PubChem CID 9589606
IUPAC Name N-[2-[(2E)-2-[1-(3-bromophenyl)ethylidene]hydrazinyl]-2-oxo-1-(4-oxo-3H-phthalazin-1-yl)ethyl]benzamide
SMILES CC(=NNC(=O)C(C1=NNC(=O)C2=CC=CC=C21)NC(=O)C3=CC=CC=C3)C4=CC(=CC=C4)Br

Specifications

Quantity 50 mg
Formula Weight 518.36
Percent Purity >97%
Chemical Name or Material AC 55541
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'