missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Access Port
Access port with silicone plug for use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600.
Brand: BINDER™ 8012-2482
This item is not returnable.
View return policy
Description
- Round access port with silicone plug
- Choose between 30, 50, or 100 mm plug
- Available on left, right, or top of the chamber
- For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, and KBF 1600
Specifications
KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600 | |
Access Port | |
50 mm |
Silicone | |
1 on Left Side |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Access Port > Access Port, 50 mm, Left
Spot an opportunity for improvement?Share a Content Correction