Learn More
BINDER™ Access Port with Silicone Plug
BINDER™ Access Port with silicone plug is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
4255.00 DKK - 7940.00 DKK
Specifications
For Use With (Equipment) | BINDER Chambers Series KBF-S and KBF-S ECO |
---|---|
Material | Steel |
Type | Access Port |
Includes | Silicone plug |
Product Code | Brand | Diameter (Metric) | Features | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Features | Price | Quantity & Availability | |||||
17333663
|
BINDER™
8012-2071 |
30 mm | Right Position |
4255.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
17343663
|
BINDER™
8012-2072 |
30 mm | Left Position |
4255.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
17353663
|
BINDER™
8012-2073 |
30 mm | Top Position |
4255.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
17363663
|
BINDER™
8012-2074 |
50 mm | Top Position |
4960.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
17373663
|
BINDER™
8012-2075 |
100 mm | Top Position |
7940.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
17383663
|
BINDER™
8012-2076 |
100 mm | Right Position |
7940.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
17393663
|
BINDER™
8012-2077 |
100 mm | Left Position |
7940.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
17303673
|
BINDER™
8012-2078 |
50 mm | Right Position |
4960.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
17313673
|
BINDER™
8012-2079 |
50 mm | Left Position |
4960.00 DKK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
Description
- Access Port
- With Silicone Plug
- For use with Constant Climate Chambers KBF-S and KBF-S ECO
Specifications
BINDER Chambers Series KBF-S and KBF-S ECO | |
Steel | |
Access Port | |
Silicone plug |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BINDER™ Access Port with Silicone Plug