Learn More
Axygen™ Accessories for Horizontal Gel Box, 15 cm
For use with horizontal gel box, 15cm
Specifications
For Use With (Equipment) Axygen Horizontal Gel Box, 15 cm
View More Specs
Products 30
Product Code Brand Type No. of Wells Thickness (Metric) Price Quantity & Availability  
Product Code Brand Type No. of Wells Thickness (Metric) Price Quantity & Availability  
15563117
View Documents
Axygen™
HGB15-10-1
Comb 10 1 mm N/A
N/A
15573117
View Documents
Axygen™
HGB15-10-15
Comb 10 1.5 mm N/A
N/A
15593117
View Documents
Axygen™
HGB15-12-1
Comb 12 1 mm N/A
N/A
15523127
View Documents
Axygen™
HGB15-14MC-15
Multi-channel Compatible Comb 14 1.5 mm N/A
N/A
15503127
View Documents
Axygen™
HGB15-12-15
Comb 12 1.5 mm N/A
N/A
15513127
View Documents
Axygen™
HGB15-14MC-1
Multi-channel Compatible Comb 14 1 mm N/A
N/A
15533127
View Documents
Axygen™
HGB15-14MC-2
Multi-channel Compatible Comb 14 2.0 mm N/A
N/A
15553127
View Documents
Axygen™
HGB15-16-1
Comb 16 1 mm N/A
N/A
15563127
View Documents
Axygen™
HGB15-16MC-1
Multi-channel Compatible Comb 16 1 mm N/A
N/A
15583127
View Documents
Axygen™
HGB15-16MC-2
Multi-channel Compatible Comb 16 2.0 mm N/A
N/A
15573127
View Documents
Axygen™
HGB15-16MC-15
Multi-channel Compatible Comb 16 1.5 mm N/A
N/A
15593127
View Documents
Axygen™
HGB15-18MC-1
Multi-channel Compatible Comb 18 1 mm N/A
N/A
15503137
View Documents
Axygen™
HGB15-18MC-15
Multi-channel Compatible Comb 18 1.5 mm N/A
N/A
15513137
View Documents
Axygen™
HGB15-20-075
Comb 20 0.75 mm N/A
N/A
15523137
View Documents
Axygen™
HGB15-20-1
Comb 20 1 mm N/A
N/A
15543137
View Documents
Axygen™
HGB15-20-2
Comb 20 2.0 mm N/A
N/A
15533137
View Documents
Axygen™
HGB15-20-15
Comb 20 1.5 mm N/A
N/A
15553137
View Documents
Axygen™
HGB15-28MC-075
Multi-channel Compatible Comb 28 0.75 mm N/A
N/A
15563137
View Documents
Axygen™
HGB15-28MC-1
Multi-channel Compatible Comb 28 1 mm N/A
N/A
15573137
View Documents
Axygen™
HGB15-28MC-15
Multi-channel Compatible Comb 28 1.5 mm N/A
N/A
15583137
View Documents
Axygen™
HGB15-30MC-1
Multi-channel Compatible Comb 30 1 mm N/A
N/A
15593137
View Documents
Axygen™
HGB15-30MC-15
Multi-channel Compatible Comb 30 1.5 mm N/A
N/A
15503147
View Documents
Axygen™
HGB15-35-15
Comb 35 1.5 mm N/A
N/A
15513147
View Documents
Axygen™
HGB15-8-1
Comb 8 1 mm N/A
N/A
15523147
View Documents
Axygen™
HGB15-8-15
Comb 8 1.5 mm N/A
N/A
15583117
View Documents
Axygen™
HGB15-10-GT
Gel Tray 15 x 10 cm, UV Transparent N/A
N/A
15543127
View Documents
Axygen™
HGB15-15-GT
Gel Tray 15 x 15 cm, UV Transparent N/A
N/A
15543147
View Documents
Axygen™
HGB15-NE
Replacement Negative Electrode, 15 cm N/A
N/A
15553147
View Documents
Axygen™
HGB15-PE
Replacement Positive Electrode, 15 cm N/A
N/A
15533147
View Documents
Axygen™
HGB15-CD
Casting Dam N/A
N/A
Specifications
Axygen Horizontal Gel Box, 15 cm