Learn More
Thermo Scientific™ Accessories for Bottle/Tube Roller
Use these accessories to optimize the performance of your Thermo Scientific™ Bottle/Tube Roller.
41.99 DKK - 884.79 DKK
Specifications
For Use With (Equipment) Bottle/Tube Roller
View More Specs
Products 5
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
15238914
View Documents Promotion Details
Thermo Scientific™
88881010
Roller
318.68 DKK
Pack of 2
In Stock
Add to basket
15258914
View Documents Promotion Details
Thermo Scientific™
88881012
O-ring for 15 mL tube
41.99 DKK
Pack of 20
In Stock
Add to basket
15248914
View Documents Promotion Details
Thermo Scientific™
88881011
O-ring for 50 mL tube
41.99 DKK
Pack of 20
15268914
View Documents Promotion Details
Thermo Scientific™
88881014
Remote Control
884.79 DKK
Each
In Stock
Add to basket
15278914
View Documents Promotion Details
Thermo Scientific™
88881013
Stacking Bar
713.83 DKK
Pack of 4
Add to basket
Description
Description
Use Thermo Scientific™ Accessories for Bottle/Tube Roller to optimize flexibility and performance.
Specifications
Specifications
Bottle/Tube Roller
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ Accessories for Bottle/Tube Roller