Learn More
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker
Optional accessories to enhance the Fisher Scientific Heating/Cooling Mini Shaker
Brand:  Fisherbrand™ 88861107
836.00 DKK valid until 2025-03-31
Use promo code "24162" to get your promotional price.
 View more versions of this product
Product Code. 15514090
  • 880.00 DKK / Each
Estimated Shipment: 29-08-2025
to see stock.
Explore more special offers
Specifications
5 x 50mL Conical Bottom Centrifuge Tube Block; Well: 29mm dia X 47.6mm depth
Special Offers
  • Fisherbrand
    Get up to 60% off select Fisherbrand products
    Promo Code: 24162
    Expires2025-03-31
    Days
    3
    Hours
    10
    Minutes
    39
    Seconds
    19
    New Offer
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions Save Now
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
    New Offer
    Save Now
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker > 5 x 50mL Conical Bottom Centrifuge Tube Block; Well: 29mm dia X 47.6mm depth