Learn More
Fisherbrand™ Accessories for Fisherbrand™ Mini Tube Rotator
Optimize your Fisher Scientific™ Mini Tube Rotator with these optional accessories
90.75 DKK
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
15524140
|
Fisherbrand™
88861201 |
Two Paddles for 10mL/15 mL tubes |
90.75 DKK
2 units |
In Stock
Log in to see stock. |
||||||
15534140
|
Fisherbrand™
88861202 |
Two Paddles for 5 mL/7mL tubes |
90.75 DKK
1 set |
In Stock
Log in to see stock. |
||||||
15544140
|
Fisherbrand™
88861203 |
Two Paddles for 1.5 mL/2mL tubes |
90.75 DKK
2 units |
In Stock
Log in to see stock. |
||||||
15554140
|
Fisherbrand™
88861204 |
Two Paddles for 0.5 mL/0.8mL tubes |
90.75 DKK
2 units |
In Stock
Log in to see stock. |
||||||
15564140
|
Fisherbrand™
88861205 |
Paddle for 50mL tubes |
90.75 DKK
Each |
Estimated Shipment: 15-10-2025
Log in to see stock. |
||||||
Description
Optimize your Mini Tube Rotator with these optional accessories
- Increase performance and flexibility
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Mini Tube Rotator