Learn More
Fisherbrand™ Accessories for Fisherbrand™ Wrist Motion Shakers
Optimize your Fisher Scientific™ Wrist Motion Shaker with these optional accessories
895.00 DKK - 4095.00 DKK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15514130
|
Fisherbrand™
88861191 |
Two 6 Position Side Arm Assembly | N/A | ||||||
15534130
|
Fisherbrand™
88861193 |
2 Vessel Clamp Assembly |
895.00 DKK
Pack of 2 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||
15504130
|
Fisherbrand™
88861190 |
Two 4 Position Side Arm Assembly |
2950.00 DKK
Pack of 2 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
|||||